Sodium hydroxide ≥27% (1.30) in aqueous solution, EMSURE® for nitrogen analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.05591.2500
1.05591.9025
CA1.05591.2500CS
790
CAD
CA1.05591.2500
CA1.05591.9025
Sodium hydroxide ≥27% (1.30) in aqueous solution, EMSURE® for nitrogen analysis, Supelco®
Sodium hydroxide
Formula:
NaOH Boiling Pt: 119 - 135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.30 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Assay (acidimetric) | ≥ 27.0 % |
Carbonate (as Na₂CO₃) | ≤ 1.0 % |
Chloride (Cl) | ≤ 0.004 % |
Phosphate (PO₄) | ≤ 0.0005 % |
Silicate (SiO₂) | ≤ 0.001 % |
Sulfate (SO₄) | ≤ 0.003 % |
Total nitrogen (N) | ≤ 0.0001 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Al (Aluminium) | ≤ 0.0005 % |
Fe (Iron) | ≤ 0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...