Sodium hydroxide ∼32% aqueous solution, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda

1.05587.2500 1.05587.9025 1.05587.9200 1.05587.5000
CA1.05587.2500CS 1823.95 CAD
CA1.05587.2500 CA1.05587.9025 CA1.05587.9200 EM1.05587.5000
Sodium hydroxide ∼32% aqueous solution, EMPLURA®, Supelco®
Sodium hydroxide

Basic cleaning agent (concentration ~32%) in premium MilliporeSigma quality. The high purity of these solutions make these sodium liquids suitable above all for cleaning applications in which residues from surfactants or complexation agents are to be avoided.


  • Strongly alkaline liquid
  • Chlorine-free


Through the use of these prepared solutions, the time-consuming and dangerous breakdown of solid sodium hydroxide can be avoided.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.35 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Assay (Acidimetric, NaOH) 32.0 - 33.0%
Carbonate (as Na₂CO₃) ≤0.5
Chloride (Cl) ≤0.004%
Phosphate (PO₄) ≤0.0005%
Silicate (SiO₂) ≤0.005%
Sulfate (SO₄) ≤0.003%
Heavy Metals (as Pb) ≤0.0005%
Al (Aluminium) ≤0.001%
Fe (Iron) ≤0.001%
Solubility (20 °C) Soluble
Boiling Point 119 °C (1013 hPa)
Vapor Pressure 8 hPa (20 °C)
pH 14 (H₂O, 20 °C)
Corrosive Material
Hazard Class Description

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR