Potassium hydroxide 47% in aqueous solution, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.05545.1000 1.05545.9025
CA1.05545.1000CS 3169.29 CAD
CA1.05545.1000 CA1.05545.9025
Potassium hydroxide 47% in aqueous solution, EMSURE® for analysis, Supelco®
Potassium hydroxide
Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.475 g/cm3 (20 °C)
pH 13.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR