Ammonia solution 25%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.05428.0250
1.05428.1000
1.05428.0500
CA1.05428.0250EA
531
CAD
CA1.05428.0250
CA1.05428.1000
CA1.05428.0500
Ammonia solution 25%, Suprapur® for trace metal analysis, Supelco®
Ammonia solution
EMD # 1.05428.0250 replaces and is identical to EMD # 5428-1.
EMD # 1.05428.1000 replaces and is identical to EMD # 5428-2.
Formula:
NH₃ Boiling Pt: 32…37.7 °C (1013 hPa) Melting Pt: –57.5 °C Density: 0.88…0.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -57.5 °C |
Boiling Point | 37.7 °C |
Vapor Pressure | 483 hPa (20 °C) |
Explosion Limit | 15.4 - 33.6 %(V) |
Density | 0.903 g/cm3 (20 °C) |
pH | (H2O, 20 °C) strongly alkaline |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...