Cresol red indicator, Supelco®

Supplier: MilliporeSigma

1.05225.0005 1.05225.0025 1.05225.0005 1.05225.0025
EM1.05225.0005CS 20339.34 CAD
EM1.05225.0005 EM1.05225.0025 CA1.05225.0005 CA1.05225.0025
Cresol red indicator, Supelco®
Cresol red
Formula: C₂₁H₁₈O₅S
Storage Temperature: Ambient
MDL Number: MFCD00005878
CAS Number: 1733-12-6
EINECS: 217-064-2

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 290 °C decomposes
Molar Mass 382.43 g/mol
Bulk Density 600 - 620 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR