Potassium sulfate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma

1.05152.0100 1.05152.0500
CA1.05152.0100EA 880 CAD
CA1.05152.0100 CA1.05152.0500
Potassium sulfate, Suprapur® for trace metal analysis, Supelco®
Potassium sulphate
Formula: K₂SO₄
Boiling Pt: 1698 °C (1013 hPa)
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Solubility 111 g/l (20 °C)
Melting Point 1069 °C
Molar Mass 174.26 g/mol
Bulk Density 800 kg/m3
Boiling Point 1689 °C (1013 hPa)
Density 2.66 g/cm3 (20 °C)
pH 5.5 - 8.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR