Potassium thiocyanate ≥99.0% (silver nitrate titration), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.05125.0250
1.05125.1000
1.05125.9050
CA1.05125.0250EA
276
CAD
CA1.05125.0250
CA1.05125.1000
CA1.05125.9050
Potassium thiocyanate ≥99.0% (silver nitrate titration), EMSURE® for analysis, Supelco®
Potassium thiocyanate
1.05125.0250 is offered as an alternative to PX1630-3. Different Pack Size, Same Specifications.
1.05125.1000 is offered as an alternative to PX1630-1. Different Pack Size, Same Specifications.
1.05125.1000 is offered as an alternative to PX1630-5. Different Pack Size, Same Specifications.
1.05125.9050 is offered as an alternative to PX1630-7. different size, different packaging, same chem spec.
1.05125.9050 replaces and is the same as PX1630-20. Part # Change only, Same Pack Size, Same Specifications.
Formula:
KSCN Boiling Pt: >400 °C (1013 hPa) (dec.) Melting Pt: 177 °C Density: 1.91 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 Merck Index: 12,07866 |
Specification Test Results
Solubility | 208 g/l (20 °C) |
Melting Point | 175 °C |
Molar Mass | 97.18 g/mol |
Bulk Density | 750 - 1000 kg/m3 |
Vapor Pressure |
|
Density | 1.89 g/cm3 (20 °C) |
pH | 5.3 - 8.5 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...