, anhydrous, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

1.05109.0100 1.05109.0500
CA1.05109.0100EA 639 CAD
CA1.05109.0100 CA1.05109.0500
, anhydrous, Suprapur® for trace metal analysis, Supelco®
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 340
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1600 g/l (20 °C)
Molar Mass 174.18 g/mol
Bulk Density 700 - 1000 kg/m3
Density 2.44 g/cm3 (20 °C)
pH 9 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR