Dipotassium hydrogen orthophosphate trihydrate ≥99.0% (by acidimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

1.05099.1000 1.05099.0250 1.05099.5000 1.05099.9050
CA1.05099.1000CS 2624 CAD
CA1.05099.1000 CA1.05099.0250 CA1.05099.5000 CA1.05099.9050
Dipotassium hydrogen orthophosphate trihydrate ≥99.0% (by acidimetry), EMSURE® for analysis, Supelco®
di-Potassium hydrogen orthophosphate trihydrate

1.05099.1000 replaces and is the same as PX1571-1. Part # Change only, Same Pack Size, Same Specifications.
1.05099.9050 replaces and is the same as PX1571-20. Part # Change only, Same Pack Size, Same Specifications.

Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ? 340 °C (1013 hPa)
Melting Pt: 340 °C (dec., anhy.)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 340 °C decomposes,(anhydrous substance)
Molar Mass 228.23 g/mol
Bulk Density 800 kg/m3
pH 9.2 - 9.4 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR