Potassium permanganate ≥99.0% (by manganometry), EMSURE® ACS for analysis (max. 0.05 ppm Hg), Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Permanganic acid potassium salt

1.05084.1000 1.05084.1000
CA1.05084.1000EA 387.76 CAD
EM1.05084.1000 CA1.05084.1000
Potassium permanganate ≥99.0% (by manganometry), EMSURE® ACS for analysis (max. 0.05 ppm Hg), Supelco®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay (manganometric) ≥ 99.0 %
Insoluble matter ≤ 0.1 %
Chloride, Chlorate (as Cl) ≤ 0.005 %
Sulfate (SO₄) ≤ 0.01 %
Total nitrogen (N) ≤ 0.003 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.002 %
Hg (Mercury) ≤ 0.000005 %
Pb (Lead) ≤ 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR