Potassium perchlorate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.05076.0250 1.05076.1000
CA1.05076.0250EA 201 CAD
CA1.05076.0250 CA1.05076.1000
Potassium perchlorate, EMSURE® for analysis, Supelco®
Potassium perchlorate
Formula: ClKO₄
Melting Pt: 608 °C
Density: 2.5239 g/cm³ (10.7 °C)
MDL Number: MFCD00011362
CAS Number: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: 5.1,II
Merck Index: 13,07738

Order Now

Specification Test Results

Solubility 17 g/l (20 °C)
Melting Point 610 °C (decomposition)
Molar Mass 138.55 g/mol
Bulk Density 1150 kg/m3
pH 5.0 (10 g/l, H2O, 25 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR