di-Potassium oxalate monohydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate

1.05073.0250 1.05073.1000
CA1.05073.0250EA 119 CAD
CA1.05073.0250 CA1.05073.1000
di-Potassium oxalate monohydrate, EMSURE® for analysis, Supelco®
di-Potassium oxalate monohydrate
Formula: (COOK)₂·H₂O
MW: 184.23 g/mol
Melting Pt: 356
Density: 2.154 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
Merck Index: 12,07820

Order Now

Specification Test Results

Solubility 360 g/l (20 °C) (anhydrous substance)
Molar Mass 184.24 g/mol
Bulk Density 700 - 1100 kg/m3
Vapor Pressure (20 °C)
Density 2.13 g/cm3 (20 °C)
pH 7.0 - 8.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR