Potassium iodate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.05051.0100 1.05051.0500
CA1.05051.0100EA 201 CAD
CA1.05051.0100 CA1.05051.0500
Potassium iodate, EMSURE® for analysis, Supelco®
Potassium iodate
Formula: KlO₃
Melting Pt: 560 °C
Density: 3.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Order Now

Specification Test Results

Solubility 92 g/l (25 °C)
Melting Point 560 °C decomposes
Molar Mass 214 g/mol
Bulk Density 2000 kg/m3
Density 3.98 g/cm3 (20 °C)
pH 6 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR