Potassium hydroxide ≥85.0% (by acidimetry), pellets, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.05033.0500 1.05033.1000 1.05033.5000 1.05033.9025 1.05033.9050
CA1.05033.0500CS 508 CAD
CA1.05033.0500 CA1.05033.1000 CA1.05033.5000 CA1.05033.9025 CA1.05033.9050
Potassium hydroxide ≥85.0% (by acidimetry), pellets, EMSURE® for analysis, Supelco®
Potassium hydroxide
Formula: KOH
Boiling Pt: 1320 °C (1013 hPa)
Melting Pt: 361 °C (lit.)
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Solubility 1130 g/l (20 °C)
Melting Point 360 °C
Molar Mass 56.11 g/mol
Boiling Point 1320 °C
Vapor Pressure (20 °C)
Density 2.04 g/cm3 (20 °C)
pH 14 (56 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR