Potassium hydroxide 84,0 % (by acidimetry), pellets, pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.05012.1000
1.05012.5000
1.05012.9050
CA1.05012.1000CS
2809.46
CAD
CA1.05012.1000
CA1.05012.5000
CA1.05012.9050
Potassium hydroxide 84,0 % (by acidimetry), pellets, pure, Supelco®
Potassium hydroxide
Formula:
KOH Boiling Pt: 1320 °C (1013 hPa) Melting Pt: 361 °C (lit.) Density: 2.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8,II REACH: 01-2119487136-33 Merck Index: 12,07806 |
Specification Test Results
Solubility | 1130 g/l (20 °C) |
Melting Point | 360 °C |
Molar Mass | 56.11 g/mol |
Boiling Point | 1320 °C |
Vapor Pressure | (20 °C) |
Density | 2.04 g/cm3 (20 °C) |
pH | 14 (56 g/l, H2O, 20 °C) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...