Potassium fluoride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.04994.0250 1.04994.1000
CA1.04994.0250EA 114 CAD
CA1.04994.0250 CA1.04994.1000
Potassium fluoride, EMSURE® for analysis, Supelco®
Potassium fluoride
Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714

Order Now

Specification Test Results

Solubility 923 g/l (20 °C)
Melting Point 860 °C
Molar Mass 58.1 g/mol
Bulk Density 400 kg/m3
Boiling Point 1500 °C (1013 hPa)
Vapor Pressure 1.3 hPa (885 °C)
Density 2.48 g/cm3 (20 °C)
pH 7 - 9 (50 g/l, H2O, 18 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR