Potassium chlorate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.04944.0100
1.04944.0500
CA1.04944.0100EA
428
CAD
CA1.04944.0100
CA1.04944.0500
Potassium chlorate, EMSURE® for analysis, Supelco®
Potassium chlorate
Formula:
KClO₃ Boiling Pt: 400 °C (1013 hPa) Melting Pt: 356 °C Density: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011361 CAS Number: 3811-04-9 EINECS: 223-289-7 UN: 1485 ADR: 5.1,II Merck Index: 13,07703 |
Specification Test Results
Solubility | 73 g/l (20 °C) |
Melting Point | 356 °C |
Molar Mass | 122.55 g/mol |
Bulk Density | 1200 - 1400 kg/m3 |
Boiling Point | 400 °C (decomposition) |
Vapor Pressure | (20 °C) |
Density | 2.32 g/cm3 (20 °C) |
pH | 5.6 (73 g/l, H2O, 20 °C) |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...