Potassium chloride ≥99.5% (silver nitrate titration), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.04936.0500 1.04936.1000 1.04936.5000 1.04936.0250 1.04936.9050
CA1.04936.0500EA 320 CAD
CA1.04936.0500 CA1.04936.1000 CA1.04936.5000 CA1.04936.0250 CA1.04936.9050
Potassium chloride ≥99.5% (silver nitrate titration), EMSURE® for analysis, Supelco®
Potassium chloride
Formula: KCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 770 °C (lit.)
Density: 1.98 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Solubility 347 g/l (20 °C)
Melting Point 773 °C
Molar Mass 74.55 g/mol
Bulk Density 1000 kg/m3
Boiling Point 1413 °C (1013 hPa)
Density 1.98 g/cm3 (20 °C)
pH 5.5 - 8.0 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR