Potassium chloride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Potassiumchloride

1.04933.0500 1.04933.0500
CA1.04933.0500EA 491.09 CAD
CA1.04933.0500 EM1.04933.0500
Potassium chloride, EMSURE® for analysis, Supelco®
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Merck Index: 13,07704

Order Now

Specification Test Results

Solubility 347 g/l (20 °C)
Melting Point 773 °C
Molar Mass 74.55 g/mol
Bulk Density 1000 kg/m3
Boiling Point 1413 °C (1013 hPa)
Density 1.98 g/cm3 (20 °C)
pH 5.5 - 8.0 (50 g/l, H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR