Potassium bromate ≥99.8%, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.04912.0100 1.04912.0250
CA1.04912.0100EA 128 CAD
CA1.04912.0100 CA1.04912.0250
Potassium bromate ≥99.8%, EMSURE® for analysis, Supelco®
Potassium bromate
Formula: KBrO₃
Melting Pt: 350 °C
Density: 3.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 12,07779

Order Now

Specification Test Results

Assay (iodometric) ≥99.8%
Insoluble matter ≤ 0.005%
pH value (5%;water;25°C) 5.0-9.0
Bromide (Br) ≤ 0.02%
Sulphate (SO₄) ≤ 0.005%
Total nitrogen (N) ≤ 0.001%
Heavy metals (as Pb) ≤ 0.0005%
Fe (Iron) ≤ 0.0005%
Na (Sodium) ≤ 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR