Potassium bromide, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.04905.0500
1.04905.0500
CA1.04905.0500CS
1120.05
CAD
CA1.04905.0500
EM1.04905.0500
Potassium bromide, EMSURE® for analysis, Supelco®
Potassium bromide
In North America 1.04905.0500 replaces but has a far superior specification to PX1380-1
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 °C (1013 hPa) Melting Pt: 730 °C Density: 2.75 g/cm³ (20 °C) |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 Merck Index: 13,07701 |
Specification Test Results
Solubility | 650 g/l (20 °C) |
Melting Point | 730 °C |
Molar Mass | 119.00 g/mol |
Bulk Density | 900 - 1000 kg/m3 |
Boiling Point | 1435 °C |
Vapor Pressure | 1.3 hPa (795 °C) |
Density | 2.75 g/cm3 (20 °C) |
pH | 5.5 - 8.5 (50 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...