Potassium hydrogen sulfate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Potassium bisulfate, Potassium bisulphate

1.04885.0500 1.04885.2500
CA1.04885.0500EA 316 CAD
CA1.04885.0500 CA1.04885.2500
Potassium hydrogen sulfate, EMSURE® for analysis, Supelco®
Potassium hydrogen sulphate
Formula: KHSO₄
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now

Specification Test Results

Solubility 490 g/l (20 °C) (development of heat)
Melting Point 195 °C (decomposition)
Molar Mass 136.17 g/mol
Bulk Density 1140 kg/m3
Density 2.24 g/cm3 (20 °C)
pH 1 (50 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR