Potassium hydrogen phthalate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: KHPPotassium phthalate monobasicPotassium Acid PhthalatePotassium biphthalatePhthalic Acid Monopotassium SaltPhthalic acid potassium salt

1.04874.1000 1.04874.1000 1.04874.0250 1.04874.0250
CA1.04874.0250EA 140.66 CAD
EM1.04874.0250 EM1.04874.1000 CA1.04874.0250 CA1.04874.1000
Potassium hydrogen phthalate, EMSURE® for analysis, Supelco®
Potassium hydrogen phthalate
Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
MDL Number: MFCD00013070
CAS Number: 877-24-7
Merck Index: 12,07773
https://ca.vwr.com/stibo/web/std.lang.all/06/58/16640658.jpg

Order Now

Specification Test Results

Solubility 80 g/l (20 °C)
Melting Point 295 - 300 °C
Molar Mass 204.22 g/mol
Bulk Density 900 kg/m3
Density 1.636 g/cm3
pH 4.0 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR