Potassium dihydrogen phosphate ≥99.5% (alkalimetric, calculated on anhydrous substance) [99.5-100.5% (alkalimetric)], EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

1.04873.0250 1.04873.1000 1.04873.5000 1.04873.9025 1.04873.9050
CA1.04873.0250EA 152 CAD
CA1.04873.0250 CA1.04873.1000 CA1.04873.5000 CA1.04873.9025 CA1.04873.9050
Potassium dihydrogen phosphate ≥99.5% (alkalimetric, calculated on anhydrous substance) [99.5-100.5% (alkalimetric)], EMSURE® for analysis, Supelco®
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Boiling Pt: ~ 100 °C (1013 hPa)
Melting Pt: 252.6 °C (lit.)
Density: 2.338 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Solubility 222 g/l (20 °C)
Melting Point 253 °C (decomposition)
Molar Mass 136.08 g/mol
Bulk Density 1200 kg/m3
Density 2.34 g/cm3 (20 °C)
pH 4.4 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR