Potassium chloride 2.9-3.1 M in aqueous solution, Supelco®

Supplier: MilliporeSigma

Synonyms: Potassiumchloride

1.04817.0250 1.04817.0250
CA1.04817.0250CS 3298.38 CAD
EM1.04817.0250 CA1.04817.0250
Potassium chloride 2.9-3.1 M in aqueous solution, Supelco®
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Density: 1.13 g/cm³ (20 °C)
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.13 g/cm3 (20 °C)
pH 5.5 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR