indicator for metal titration, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid dihydrate, 2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalene-3-carboxylic acid dihydrate

M-Clarity™ quality level = MQ200

1.04595.0005 1.04595.0025 1.04595.0005 1.04595.0025
EM1.04595.0005CS 26452.36 CAD
EM1.04595.0005 EM1.04595.0025 CA1.04595.0005 CA1.04595.0025
indicator for metal titration, Supelco®
Patton-reeders reagent (Calconcarboxylic acid dihydrate)
Formula: C₂₁H₁₄N₂O₇S·2H₂O
MW: 474.45 g/mol
MDL Number: MFCD00004078
CAS Number: 3737-95-9
EINECS: 223-117-0

Order Now

Specification Test Results

Identity (UV/VIS-Spectrum) passes test
Appearance of solution (0.1 g/l; ethanol) passes test
Absorption maximum λmax. (Ethanol) 569 - 572 nm
Spec. Absorptivity A 1%/1cm (λmax.; 0.025 g/l; ethanol; calc. on water free substance) ≥ 250
Water (according to Karl Fischer) ≤ 9 %
Suitability as indicator (for metal titration) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR