Fluorescein disodium salt indicator (C.I. 45350), Supelco®

Supplier: MilliporeSigma

1.03887.0050 1.03887.0250 1.03887.0050 1.03887.0250
EM1.03887.0050CS 2376.32 CAD
EM1.03887.0050 EM1.03887.0250 CA1.03887.0050 CA1.03887.0250
Fluorescein disodium salt indicator (C.I. 45350), Supelco®
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
Melting Pt: > 300 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 300 °C
Molar Mass 376.27 g/mol
Bulk Density 600 kg/m3
pH 8.3 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR