Iron(III) nitrate nonahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

M-Clarity™ quality level = MQ300

1.03883.0250 1.03883.1000
CA1.03883.0250EA 132 CAD
CA1.03883.0250 CA1.03883.1000
Iron(III) nitrate nonahydrate, EMSURE® for analysis, Supelco®
Iron (III) nitrate nonahydrate

1.03883.0250 replaces and is the same as FX0225-1. Different Pack Size, Same Specifications. 1.03883.1000 replaces and is the same as FX0225-4. Part # Change only, Same Pack Size, Same Specifications. 1.03883.1000 replaces and is the same as FX0225-5. Different Pack Size, Same Specifications. 1.03883.1000 replaces and is the same as FX0225-7. Different Pack Size, Same Specifications. 1.03883.9050 replaces and is the same as FX0225-7. Different Pack Size, Same Specifications.

Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125
Melting Pt: 47
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 8 II
Merck Index: 13,04057

Order Now

Specification Test Results

Assay (complexometric) 99.0 - 101.0 %
Insoluble matter ≤ 0.005 %
Free acid (as HNO₃) ≤ 0.3 %
Chloride (Cl) ≤ 0.0005 %
Phosphate (PO₄) ≤ 0.005 %
Sulfate (SO₄) ≤ 0.005 %
Ca (Calcium) ≤ 0.005 %
Cu (Copper) ≤ 0.005 %
K (Potassium) ≤ 0.005 %
Mg (Magnesium) ≤ 0.001 %
Mn (Manganese) ≤ 0.02 %
Na (Sodium) ≤ 0.005 %
Pb (Lead) ≤ 0.001 %
Zn (Zinc) ≤ 0.001 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR