Ammonium iron (II) sulfate hexahydrate, EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

MQ300

1.03792.0500 1.03792.1000 1.03792.5000
CA1.03792.0500EA 513 CAD
CA1.03792.0500 CA1.03792.1000 CA1.03792.5000
Ammonium iron (II) sulfate hexahydrate, EMSURE® analysis_for, Supelco®
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 100
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Assay (manganometric) 99.0 - 101.5 %
pH-value (5 %; water) 3.0 - 5.0
Chloride (Cl) ≤ 0.001 %
Phosphate (PO₄) ≤ 0.002 %
Iron(III)-salts (Fe³⁺) ≤ 0.02 %
Ca (Calcium) ≤ 0.002 %
Cu (Copper) ≤ 0.002 %
K (Potassium) ≤ 0.01 %
Mg (Magnesium) ≤ 0.01 %
Mn (Manganese) ≤ 0.05 %
Na (Sodium) ≤ 0.01 %
Pb (Lead) ≤ 0.001 %
Zn (Zinc) ≤ 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR