Bromothymol blue indicator, Supelco®

Supplier: MilliporeSigma

1.03026.0005 1.03026.0025 1.03026.0005 1.03026.0025
EM1.03026.0005CS 32598.56 CAD
EM1.03026.0005 EM1.03026.0025 CA1.03026.0005 CA1.03026.0025
Bromothymol blue indicator, Supelco®
Bromothymol blue
Formula: C₂₇H₂₈Br₂O₅S
Melting Pt: 204…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 624.38 g/mol
Bulk Density 450 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR