Copper(II) sulfate, anhydrous, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Cupric sulphate, Cupric sulfate

1.02791.0250 1.02791.1000
CA1.02791.0250EA 216 CAD
CA1.02791.0250 CA1.02791.1000
Copper(II) sulfate, anhydrous, EMSURE® for analysis, Supelco®
Copper (II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200
Density: 3.603 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3077
ADR: 9 III
Merck Index: 12,02722

Order Now

Specification Test Results

Solubility 203 g/l (20 °C)
Molar Mass 159.61 g/mol
Bulk Density 800 kg/m3
Density 3.60 g/cm3 (20 °C)
pH 3.5 - 4.5 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR