Copper(II) nitrate trihydrate ≥99.5% (by iodometry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate

1.02753.0250 1.02753.1000 1.02753.9025
CA1.02753.0250EA 114 CAD
CA1.02753.0250 CA1.02753.1000 CA1.02753.9025
Copper(II) nitrate trihydrate ≥99.5% (by iodometry), EMSURE® for analysis, Supelco®
Copper (II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Melting Pt: 114 °C
Density: 2.05 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 10031-43-3
EINECS: 221-838-5
UN: 1477
ADR: 5.1,II
Merck Index: 13,02671

Order Now

Specification Test Results

Solubility 2670 g/l (20 °C)
Melting Point 114 °C
Molar Mass 241.60 g/mol
Bulk Density 1050 kg/m3
Density 2.05 g/cm3 (20 °C)
pH 3 - 4 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR