Cobalt(II) sulfate heptahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02556.0100
1.02556.0250
CA1.02556.0100EA
544
CAD
CA1.02556.0100
CA1.02556.0250
Cobalt(II) sulfate heptahydrate, EMSURE® for analysis, Supelco®
Cobalt (II) sulphate heptahydrate
Formula:
CoSO₄·7H₂O Melting Pt: 735 °C Density: 2.03 g/cm³ (20 °C) |
MDL Number:
MFCD00149658 CAS Number: 10026-24-1 EINECS: 233-334-2 UN: 3077 ADR: 9,III Merck Index: 12,02510 |
Specification Test Results
Solubility | 362 g/l (20 °C) |
Melting Point | 98 °C |
Molar Mass | 281.10 g/mol |
Bulk Density | 800 - 1100 kg/m3 |
Density | 1.95 g/cm3 (20 °C) |
pH | 4 (100 g/l, H2O, 20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...