Cobalt(II) chloride hexahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

1.02539.0100 1.02539.0250
CA1.025390100CS 32751.36 CAD
CA1.025390100 CA1.025390250
Cobalt(II) chloride hexahydrate, EMSURE® for analysis, Supelco®
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 56 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now

Specification Test Results

Solubility 191 g/l (100 °C)
Melting Point 56 °C
Molar Mass 237.90 g/mol
Bulk Density 1250 kg/m3
Density 1.92 g/cm3 (25 °C)
pH 4.9 (50 g/l, H2O, 25 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR