Cobalt(II) nitrate hexahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02536.0100
1.02536.0250
CA1.02536.0100EA
546
CAD
CA1.02536.0100
CA1.02536.0250
Cobalt(II) nitrate hexahydrate, EMSURE® for analysis, Supelco®
Cobalt (II) nitrate hexahydrate
Formula:
Co(NO₃)₂·6H₂O Melting Pt: 57 °C Density: 1.88 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149647 CAS Number: 10026-22-9 EINECS: 233-402-1 UN: 3077 ADR: 9,III Merck Index: 13,02469 |
Specification Test Results
Solubility | 2170 g/l (100 °C) |
Melting Point | 57 °C |
Molar Mass | 291.04 g/mol |
Bulk Density | 800 kg/m3 |
Density | 1.87 g/cm3 (20 °C) |
pH | 4.0 (100 g/l, H2O, 20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...