Sodium hexanitritocobaltate(III), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium cobaltnitrite, Trisodium hexanitritocobaltate

1.02521.0025 1.02521.0100
CA1.02521.0025EA 184 CAD
CA1.02521.0025 CA1.02521.0100
Sodium hexanitritocobaltate(III), EMSURE® for analysis, Supelco®
Sodium hexanitritocobaltate(III)
Formula: Na₃Co(NO₂)₆
MW: 403.94 g/mol
Melting Pt: ∼220 °C
Storage Temperature: Ambient
MDL Number: MFCD00003512
CAS Number: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1 III

Order Now

Specification Test Results

Solubility 720 g/l (20 °C)
Melting Point 220 °C
Molar Mass 403.93 g/mol
Bulk Density 480 kg/m3
Density 0.48 g/cm3
pH 5 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR