Calcium carbonate, Certipur® volumetric standard, secondary reference material, traceable to NIST Standard Reference Material (SRM), Supelco®
Supplier: MilliporeSigma
Synonyms:
Marble
1.02410.0050
CA1.02410.0050EA
140.11
CAD
CA1.02410.0050
Calcium carbonate, Certipur® volumetric standard, secondary reference material, traceable to NIST Standard Reference Material (SRM), Supelco®
Calcium carbonate
Formula:
CaCO₃ MW: 100.09 g/mol Melting Pt: 825 °C Density: 2.71 g/cm³ (20 °C) |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 Merck Index: 13,01658 |
Specification Test Results
Solubility | 0.014 g/l (20 °C) |
Melting Point | 825 °C (decomposition) |
Molar Mass | 100.09 g/mol |
Bulk Density | 400 - 1400 kg/m3 |
pH | 9.5 - 10.5 (100 g/l, H2O, 20 °C) (slurry) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...