Cerium(IV) sulfate tetrahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02274.0025
1.02274.0100
1.02274.0250
CA1.02274.0025EA
250
CAD
CA1.02274.0025
CA1.02274.0100
CA1.02274.0250
Cerium(IV) sulfate tetrahydrate, EMSURE® for analysis, Supelco®
Cerium (IV) sulphate tetrahydrate
Formula:
Ce(SO₄)₂·4H₂O Melting Pt: 180…200 °C Density: 3.91 g/cm³ (20 °C) |
MDL Number:
MFCD00149427 CAS Number: 10294-42-5 EINECS: 237-029-5 UN: 3260 ADR: 8,II Merck Index: 12,02036 |
Specification Test Results
Solubility | 38 g/l (50 °C) |
Melting Point | 180 - 200 °C Elimination of water of crystallisation |
Molar Mass | 404.30 g/mol |
Bulk Density | 650 - 850 kg/m3 |
Density | 5.02 g/cm3 (20 °C) |
pH | 1.6 (10 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...