Calcium carbonate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma

1.02059.0050 1.02059.0500
CA1.02059.0050EA 479 CAD
CA1.02059.0050 CA1.02059.0500
Calcium carbonate, Suprapur® for trace metal analysis, Supelco®
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now

Specification Test Results

Solubility 0.014 g/l (20 °C)
Melting Point 825 °C (decomposition)
Molar Mass 100.09 g/mol
Bulk Density 400 - 1400 kg/m3
pH 9.5 - 10.5 (100 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR