Cesium chloride, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.02039.0050
1.02039.0250
1.02039.1000
CA1.02039.0050CS
182668.87
CAD
CA1.02039.0050
CA1.02039.0250
CA1.02039.1000
Cesium chloride, Suprapur® for trace metal analysis, Supelco®
Cesium chloride
Formula:
CsCl Boiling Pt: 1303 °C (1013 hPa) Melting Pt: 646 °C Density: 3.7808 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010955 CAS Number: 7647-17-8 EINECS: 231-600-2 Merck Index: 13,02021 |
Specification Test Results
Solubility | 1860 g/l (20 °C) |
Melting Point | 646 °C |
Molar Mass | 168.36 g/mol |
Bulk Density | 1800 kg/m3 |
Boiling Point | 1382 °C |
Density | 3.97 g/cm3 (20 °C) |
pH | 7 - 9 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...