Dragendorff's reagent, spray reagent for TLC, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02035.0100
1.02035.0100
EM1.02035.0100EA
303.01
CAD
EM1.02035.0100
CA1.02035.0100
Dragendorff's reagent, spray reagent for TLC, Supelco®
Dragendorff's reagent
Boiling Pt:
77 °C (1013 hPa) Density: 1.2 g/cm³ (20 °C) Flash Pt: –4 °C |
MDL Number:
MFCD00283106 CAS Number: 39775-75-2 UN: 2924 ADR: 3 (8),II |
Specification Test Results
Boiling Point | 77 °C |
Density | 0.95 g/cm3 (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...