Barium perchlorate, anhydrous, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01738.0250 1.01738.1000
CA1.01738.0250EA 409 CAD
CA1.01738.0250 CA1.01738.1000
Barium perchlorate, anhydrous, EMSURE® for analysis, Supelco®
Barium perchlorate
Formula: Ba(ClO₄)₂
MW: 336.23 g/mol
Melting Pt: 505
Density: 3.2 g/cm³ (20 °C)
Flash Pt: 21 °C (closed cup)
MDL Number: MFCD00003440
CAS Number: 13465-95-7
EINECS: 236-710-4
UN: 1447
ADR: 5.1 (6.1) II
Merck Index: 12,01016

Order Now

Specification Test Results

Solubility 1985 g/l (25 °C)
Melting Point 505 °C
Molar Mass 336.23 g/mol
Bulk Density 1300 kg/m3
Density 2.74 g/cm3 (20 °C)
pH 5.6 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR