Barium chloride dihydrate ≥99.0% (by complexometric analysis), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01719.0500 1.01719.1000 1.01719.5000 1.01719.9050
CA1.01719.0500EA 172 CAD
CA1.01719.0500 CA1.01719.1000 CA1.01719.5000 CA1.01719.9050
Barium chloride dihydrate ≥99.0% (by complexometric analysis), EMSURE® for analysis, Supelco®
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3.86 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Solubility 358 g/l (20 °C) (anhydrous substance)
Melting Point 963 °C (anhydrous substance)
Molar Mass 244.28 g/mol
Bulk Density 1200 - 1400 kg/m3
Boiling Point 1560 °C (anhydrous substance)
Density 3.86 g/cm3
pH 5.2 - 8.0 (50 g/l, H2O, 25 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR