Barium chloride, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.01716.0050 1.01716.0500
CA1.01716.0050CS 31644 CAD
CA1.01716.0050 CA1.01716.0500
Barium chloride, Suprapur® for trace metal analysis, Supelco®
Barium chloride
Formula: BaCl₂
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: ∼963 °C
Density: 2.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998

Order Now

Specification Test Results

Solubility 375 g/l (20 °C)
Melting Point 963 °C
Molar Mass 208.23 g/mol
Bulk Density 1350 kg/m3
Boiling Point 1560 °C
Density 3.9 g/cm3 (20 °C)
pH 5 - 8 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR