Barium carbonate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.01714.0250 1.01714.1000
CA1.01714.0250EA 286 CAD
CA1.01714.0250 CA1.01714.1000
Barium carbonate, EMSURE® for analysis, Supelco®
Barium carbonate
Formula: BaCO₃
Melting Pt: 1350 °C
Density: 3.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972

Order Now

Specification Test Results

Solubility 0.02 g/l (20 °C)
Melting Point 1450 °C (decomposition)
Molar Mass 197.34 g/mol
Bulk Density 350 kg/m3
Density 4.43 g/cm3 (20 °C)
pH 7 - 8 (0.016 g/l, H2O, 16 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR