Silver nitrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

1.01512.0025 1.01512.0100 1.01512.0250 1.01512.1000
CA1.01512.0025CS 632974.91 CAD
CA1.01512.0025 CA1.01512.0100 CA1.01512.0250 CA1.01512.1000
Silver nitrate, EMSURE® for analysis, Supelco®
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 2160 g/l (20 °C)
Melting Point 212 °C
Molar Mass 169.87 g/mol
Bulk Density 2350 kg/m3
Boiling Point 444 °C (1013 hPa) (decomposition)
Density 4.35 g/cm3 (20 °C)
pH 5.4 - 6.4 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR