Neutral red indicator, for microbiology (C.I. 50040), Supelco®

Supplier: MilliporeSigma

Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5

1.01369.0025 1.01369.0100 1.01369.0025 1.01369.0100
EM1.01369.0025CS 9884.48 CAD
EM1.01369.0025 EM1.01369.0100 CA1.01369.0025 CA1.01369.0100
Neutral red indicator, for microbiology (C.I. 50040), Supelco®
Neutral red
Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571

Order Now

Specification Test Results

Solubility 50 g/l (25 °C)
Molar Mass 288.78 g/mol
Bulk Density 350 - 500 kg/m3
pH 3.1 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR