Methyl orange indicator (C.I. 13025), Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

1.01322.0025 1.01322.0100 1.01322.1000 1.01322.0025 1.01322.0100 1.01322.1000
EM1.01322.0025CS 3992.66 CAD
EM1.01322.0025 EM1.01322.0100 EM1.01322.1000 CA1.01322.0025 CA1.01322.0100 CA1.01322.1000
Methyl orange indicator (C.I. 13025), Supelco®
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: > 300 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: 38
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1 III
Merck Index: 13,06127

Order Now

Specification Test Results

Solubility 5 g/l (20 °C)
Melting Point 300 °C
Molar Mass 327.34 g/mol
Bulk Density 200 - 400 kg/m3
pH 6.5 (5 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR