Ammonium sulfate ≥99.5% (by alkalimetry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
1.01217.1000
1.01217.0100
1.01217.5000
1.01217.9025
CA1.01217.1000CS
1258
CAD
CA1.01217.1000
CA1.01217.0100
CA1.01217.5000
CA1.01217.9025
CA1.01217.9050
Ammonium sulfate ≥99.5% (by alkalimetry), EMSURE® for analysis, Supelco®
Ammonium sulphate
Formula:
(NH₄)₂SO₄ Melting Pt: >280 °C (dec., lit.) Density: 1.77 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Specification Test Results
Solubility | 754 g/l (20 °C) |
Molar Mass | 132.14 g/mol |
Bulk Density | 850 kg/m3 |
Density | 1.77 g/cm3 (20 °C) |
pH | 5 (100 g/l, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...