Ammonium sulfate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma

1.01209.0050 1.01209.0500
CA1.01209.0050CS 175608.8 CAD
CA1.01209.0050 CA1.01209.0500
Ammonium sulfate, Suprapur® for trace metal analysis, Supelco®
Ammonium sulphate
Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Solubility 754 g/l (20 °C)
Molar Mass 132.14 g/mol
Bulk Density 850 kg/m3
Density 1.77 g/cm3 (20 °C)
pH 5 (100 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR